Learn More
NAlpha, NEpsilon-Bis-boc-L-lysine, TRC
Codice prodotto. 30240579
Cambia vista
Click to view available options
Quantità:
250 mg
1 g
5 g
Dimensione della confezione:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30240579 5 g 5g
30240533 250 mg 250mg
30240892 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 30240579 Fornitore Toronto Research Chemicals N. del fornitore TRCB6569955G
 Articolo soggetto a restrizioni
Spedizione stimata: 13-08-2026
Aggiungi al carrello
Aggiungi al carrello
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 2483-46-7
Formula molecolare C16 H30 N2 O6
Molecular Weight (g/mol) 346.42
Sinonimo L-Lysine, N2,N6-bis[(1,1-dimethylethoxy)carbonyl]-, Lysine, N2,N6-dicarboxy-, N2,N6-di-tert-butyl ester (7CI), Lysine, N2,N6-dicarboxy-, N2,N6-di-tert-butyl ester, L- (8CI), N2,N6-Bis[(1,1-dimethylethoxy)carbonyl]-L-lysine, (S)-2,6-Bis(tert-butoxycarbonylamino)hexanoic acid, (S)-2,6-Bis(tert-butyloxycarbonylamino)hexanoic acid, Di(N-Boc)-L-lysine, Di-tert-butoxycarbonyl-L-lysine, Di[N-(tert-Butoxycarbonyl)]-L-lysine, N,N'-Bis(tert-butoxycarbonyl)-(S)-lysine, N,N'-Bis-tert-butoxycarbonyl-L-lysine, N,N'-Di-tert-butoxycarbonyl-L-lysine, N2,N6-Bis(tert-butoxycarbonyl)-L-lysine, NSC 164052
IUPAC Name (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)O
Specifica
Materiale o nome chimico Na, Ne-Bis-boc-L-lysine
InChI Formula InChI=1S/C16H30N2O6/c1-15(2,3)23-13(21)17-10-8-7-9-11(12(19)20)18-14(22)24-16(4,5)6/h11H,7-10H2,1-6H3,(H,17,21)(H,18,22)(H,19,20)/t11-/m0/s1
Quantità 5 g
Peso formulazione 346.2104
Shipping Condition Room Temperature