Descrizione
Easy and fast measurements:
- Using bar coded test tubes, the measurement is automatically performed after inserting the cuvette into the cuvette slot.
- Contact with any dangerous chemicals is avoided because the measurement is performed directly in the test tube.
- This guarantees maximum safety for the user.
Approved:
- Reliable and comparable results.
- The reaction basis of tests is based on internationally approved standard methods including DIN-, EN-, ISO-, EPA, and APHA.
Versatile:
- The perfect test for every request.
- A large number of tests with different ranges is available for all important parameters in water and wastewater analysis.
Precise:
- Easy handling and highest accuracy.
- All tests contain accurate predosed reagents that are ready to use.
Unique:
- No mix up of tests.
- Every test box is equipped with a color-coded label and can be clearly identified.
Specifica
Specifica
| Analiti | Sulfide 3 |
| Descrizione | Seawater suitable after dilution (1 +3) |
| N. di test | 20 Determinations |
| Range | 0.05 to 3.00 mg/L |
| Metodo test | DIN method: N,N-Dimethyl-1,4-phenylene diamine reacts with hydrogen sulphide to form a compound which changes to leucomethylene blue. This compound is oxidized to methylene blue using iron(III) ions (Reaction basis according to DIN 38405-D26/27) |
| Tipo | Nanocolor™ PhotometricTube Test |