Description
Certified standards for verifying instrument performance to factory specifications are available separately.
This kit is useful for reconditioning the sample measurement pedestals to a hydrophobic state if they become “unconditioned” (see user manual for more information). The PR-1 kit consists of a small screw cap vial of specially formulated compound and a supply of 25 convenient applicators. Kit sold separately from instrument.
For use with Thermo Scientific™ NanoDrop™ Spectrophotometers.
For use with Thermo Scientific™ NanoDrop™ Spectrophotometers.
Includes:
Small screw cap vial of specially formulated reconditioning compound and 25 convenient swabs
Specifications
Specifications
| Tipo | Kit di ricondizionamento NanoDrop PR-1 |
| Da utilizzare con (apparecchiatura) | Tutti gli spettrofotometri Thermo Scientific NanoDrop |