missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NanoDrop™ Ultra FL-1 Fluorescence Verification Kit

Codice prodotto. 31921049 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
1 Kit
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
31921049 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 31921049 Fornitore Thermo Scientific™ N. del fornitore CHEMFL1KIT
 Articolo soggetto a restrizioni

Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.

Richiedi qui il prezzo per questo articolo.

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The NanoDrop Ultra FL-1 Fluorescence Verification Assay Kit is essential for verifying the fluorometer performance in NanoDrop Ultra FL and NanoDrop UltraC FL spectrophotometers. It includes all necessary reagents: Blank Reagent, Green Fluorescence Reagent, and Far Red Fluorescence Reagent.

The Thermo Scientific NanoDrop Ultra FL-1 Fluorescence Verification Kit is a specialized assay system designed to ensure optimal performance verification of fluorometer components in NanoDrop Ultra FL and UltraC FL spectrophotometers. This comprehensive kit provides essential reagents, including Blank Reagent, Green Fluorescence Reagent, and Far Red Fluorescence Reagent.

Engineered specifically for researchers conducting RNA analyses through qPCR and RT-PCR methodologies, the NanoDrop Ultra FL-1 Fluorescence Verification Kit supports the advanced microliter multimode detection capabilities of NanoDrop Ultra/UltraC FL instruments. The integration of robust fluorescence assay verification ensures streamlined workflows while maintaining effortless compliance with 21 CFR Part 11 regulatory requirements.

Features of the NanoDrop Ultra FL-1 Fluorescence Verification Kit include:

  • Comprehensive reagent kit—contains three essential reagents, including Blank Reagent, Green Fluorescence Reagent, and Far Red Fluorescence Reagent for complete system calibration and performance verification
  • Instrument-specific compatibility—engineered exclusively for NanoDrop Ultra FL and UltraC FL spectrofluorometer models to ensure optimal system integration and measurement accuracy
  • User-friendly implementation—supplied with comprehensive setup and operational instructions to facilitate proper reagent handling and calibration procedures
  • Preventive maintenance protocol—designed for biannual application (every six months) to maintain instrument precision and extend operational lifespan
  • Multi-wavelength calibration capability—enables calibration across multiple fluorescence detection channels through green and far-red fluorescent standards
  • Quality assurance standards—provides blank reference material for baseline correction and systematic error identification during routine instrument validation

Laboratory quality assurance and measurement integrity

By implementing the NanoDrop Ultra FL-1 Fluorescence Verification Assay Kit, laboratories establish a robust quality assurance framework that ensures their NanoDrop Ultra FL and UltraC FL devices maintain optimal functional performance. This systematic approach to instrument validation directly supports the maintenance of measurement integrity across all fluorescence-based analytical procedures. The verification process enables laboratories to demonstrate compliance with quality standards while providing documented evidence of instrument performance, thereby supporting regulatory requirements and internal quality management systems.

Warranty

3 Year Warranty

Specifica

Saggio Fluorescence Verification
Content And Storage 2°C to 8°C
Metodo di rilevamento Fluorescence
Formato Kit
Da utilizzare con (apparecchiatura) NanoDrop Ultra
N. di reazioni 25 Assays
Product Type Verification Kit
Quantità 1 Kit
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.