missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naproxen-d3, TRC
Specifica
Specifica
| Materiale o nome chimico | Naproxen-d3 |
| CAS | 958293-79-3 |
| Note sul grado di purezza | HPLC |
| Formula molecolare | C14H11D3O3 |
| Quantità | 50 mg |
| Sinonimo | 2-Naphthaleneacetic acid','6-(methoxy-d3)-α-methyl-, 6-(Methoxy-d3)-α-methyl-2-naphthaleneacetic acid, 2-[6-(Trideuteromethoxy)naphthalen-2-yl]propanoic acid, rac-Naproxen (methoxy-D3), (R,S)-Naproxen D3 (methoxy D3) |
| InChI Formula | InChI=1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/i2D3 |
| SMILES | [2H]C([2H])([2H])Oc1ccc2cc(ccc2c1)C(C)C(=O)O |
| IUPAC Name | 2-[6-(trideuteriomethoxy)naphthalen-2-yl]propanoic acid |
| Molecular Weight (g/mol) | 233.28 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?