missing translation for 'onlineSavingsMsg'
Learn More

Ark Plas Products™ Connettori riduttori scanalati Premium in Kynar naturale

Use these fittings for a wide range of applications. Ark Plas Products™ Natural Kynar Premium Grade Barbed Reducing Connectors are molded in lot-traceable, virgin materials for critical use applications.

Marca:  Ark Plas Products™ MAD39-DF0

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 11851140

  • 37.34€ / pezzi 10


Spedizione stimata
27-07-2021
Aggiungi al carrello

Descrizione

Descrizione

Premium fittings are stocked in White Nylon, Natural Polypropylene, Clear Polycarbonate and Kynar (PVDF). Available in two barb styles:

The Standard Barb
  • Designed to accommodate most types of flexible tubing
  • Inner diameter can expand 40% to 50% above nominal ID value
  • Holds up to 125 PSI without the use of tubing clamps
The Hard-Line Barb
  • Designed for use with semi-rigid tubing
  • Best suited for low-pressure flexible tubing applications
  • Tubing is expanded 21% above nominal ID value
Specifica

Specifica

Naturale
Barbed Reducing Connector
1/4 x 5/32 in. Barb
Kynar
Confezione da 10
Videos
SDS
Documenti