Specifica
| Concentrazione | 1 μg/μL |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Quantità | 0.1 mg |
| Tipo | Negative Control Oligodeoxynucleotide |