Learn More
Lovibond™ Tintometer™ Nessler Tube with Plungers
Product Code. 13151671
Change view
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
13151671 50 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13151671 Supplier Lovibond™ Supplier No. 353080
Spedizione stimata: 10-12-2026
Used to assess product colour by comparison with representative series of fixed colour standards
  • Anti-meniscus plungers
Specifications
Volume (Metric) 50 mL
Length (Metric) 113 mm
For Use With (Equipment) Nessleriser System 2150
Quantity 2 Pack