Learn More
Thermo Scientific™ Neuropeptide Y (13-36), Human
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| Specie | Human |
| Metodo di purificazione | HPLC |
| Content And Storage | -30°C to -10°C |
| Formato | Solid |
| Peso molecolare | 3000.43 |
| Product Type | Neuropeptide Y (13-36), human |
| Quantità | 0.5 mg |
| Status giuridico | RUO - research use only |
| Sequenza | Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2 |
For Research Use Only