Learn More
Nicotine [for single use], LoGiCal
Product Code. 30457562
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457562 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30457562 Supplier LoGiCal Supplier No. LGCFOR0517.00100
 Restricted Item
Spedizione stimata: 30-11-2026
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Nicotine
CAS 54-11-5
Analyte or Component Names Nicotine
Shipping Condition Ice pack (-20°C)
Molecular Formula C10 H14 N2
Certifications/Compliance ISO 9001
Synonym Nicotine, (-)-Nicotine, (-)-(S)-Nicotine, (-)-3-(1-Methyl-2-pyrrolidyl)pyridine, (S)-(-)-Nicotine, (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine, (S)-Nicotine, 3-[(2S)-1-Methyl-2-pyrrolidinyl]pyridine, Exodus, Flux Maag, Habitrol, L-Nicotine, NSC 5065, Nicabate, Nicoderm, Nicolan, Niconil, Nicopatch, Nicorette, Nicotell TTS, Nicotin, Nicotinell, Nicotrol, Nikofrenon, Niquitin, Tabazur, XL All Insecticide, l-Nicotine, Nicotine (8CI), Pyridine, 3-(1-methyl-2-pyrrolidinyl)-, (S)-, (-)-beta-Pyridyl-alpha-N-methylpyrrolidine
InChI Formula InChI=1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-/m0/s1
Solution Type Neat
SMILES CN1CCC[C@H]1c2cccnc2
IUPAC Name 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Industry Type Clinical Testing
Molecular Weight (g/mol) 162.23
Quantity 100 mg
Formula Weight 162.1157 g/mol