missing translation for 'onlineSavingsMsg'
Learn More

Niton™ XL2 XRF Precious Metal Analyzer

Codice prodotto. 18883468 Sfoglia Tutto Thermo Scientific Prodotti
Change view
Click to view available options
Tipo di rilevatore:
High-performance Semiconductor
Tipo di provetta:
Ag anode 45 kV maximum, 80μA maximum
Tipo di display:
Fixed angle, color, touch-screen display
Unit Size:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tipo di rilevatore Tipo di provetta Tipo di display Unit Size
18883468 High-performance Semiconductor Ag anode 45 kV maximum, 80μA maximum Fixed angle, color, touch-screen display Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18883468 Supplier Thermo Scientific Supplier No. XL2PRECIOUS
 Articolo soggetto a restrizioni

Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.

Richiedi qui il prezzo per questo articolo.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform accurate gold analysis, quickly detect gold plating and counterfeit gold and test the purity and chemistry of all precious metals with handheld XRF analyzers.

With the volatility and high price of gold, failing to detect just a small variation in composition can be an expensive mistake. The Thermo Scientific™ Niton™ XL2 XRF Precious Metal Analyzer provides a fast, accurate, and most importantly, nondestructive method to test the purity and chemistry of all precious metals. The Niton XL2 XRF Precious Metal Analyzer quickly detects gold plating and counterfeit gold and gives you the exact elemental composition of jewelry and coins so you can be absolutely certain of the value of the precious metals you buy and sell.

The Niton XL2 XRF Precious Metal Analyzer delivers accurate analysis in just seconds in an easy-to-use, robust and reliable package. Measure the content of all gold and precious metals with no need to manually change your calibration based on the metal you're analyzing—e.g., gold (Au), silver (Ag), platinum (Pt), palladium (Pd).

  • Easier, faster and more accurate than nitric acid test methods.
  • Faster, more comprehensive analysis than fire assay with comparable accuracy.
  • Efficiently identify most gold-plated items with Thermo Scientific AuDIT™ gold-plating detection technology.
  • Precisely determine the presence and concentration of other trace alloying elements and dangerous heavy elements, which could impact valuation and future refining needs.

Features:

  • Standard analytical range: 22 elements from S to U (varies by application).
  • Point and shoot simplicity—very easy to use even by nontechnical personnel.
  • Ideally suited for retail environments.
  • Nondestructive analysis with near-instantaneous results.
  • Ergonomic design.
TRUSTED_SUSTAINABILITY

Specifications

Applicazioni Non-destructive method to test the purity and chemistry of precious metals
Modalità analitiche disponibili Varies by application. Alloy modes: metal alloy, electronics alloy, precious metals. Bulk modes: mining, soil. Plastic modes: RoHS plastics, toy and consumer goods plastics, Thermo Scientific TestAll, painted products. Custom modes: upon request
Memoria dati 64MB internal system memory/128MB internal user storage
Memorizzazione dei dati Internal >10,000 readings with spectra
Trasferimento dati USB, Bluetooth and RS-232 serial communication
Descrizione Niton XL2 XRF Precious Metal Analyzer
Tipo di rilevatore High-performance Semiconductor
Tipo di display Fixed angle, color, touch-screen display
Altezza (sistema imperiale) 4 in.
Altezza (sistema metrico) 100 mm
Lunghezza (sistema imperiale) 10.25 in.
Lunghezza (sistema metrico) 256 mm
Larghezza (sistema imperiale) 11 in.
Larghezza (sistema metrico) 275 mm
Accessori opzionali Portable test stand; Stationary (bench-top) stand; Mobile test stand; Welding mask; Soil testing guard
Consumo energetico 100μA
Funzioni di sicurezza Password-protected user security
Accessori standard Locking shielded carrying case; Shielded belt holster; Two 6-cell lithium-ion battery packs; 110/220VAC battery charger/AC adaptor; PC connection cables (USB and RS-232); Thermo Scientific™ Niton Data Transfer (NDT™) PC software; Safety lanyard; Check samples/standards
Dettagli sistema 4096 channel MCA; 64MB internal system memory/128MB internal user storage
Tipo di provetta Ag anode 45 kV maximum, 80μA maximum
Tensione del tubo 45 kV
Peso (sistema imperiale) 3.0 lb., 5.8 oz.
Peso (sistema metrico) 1.53 kg
Elettronica del sistema 400 MHz ARM 11 CPU, 300 MHz dedicated DSP, 80 MHz ASICS DSP for signal processing
Immissione dati Touch-screen keyboard, user-programmable pick lists, optional wireless remote barcode reader
Licensing Registration: Varies by region. Contact your local distributor.
Dimensioni (L x P x A) 10.25 x 11 x 4 in. (256 x 275 x 100 mm)
Sistema operativo Ownership
Certificazione/Conformità CE, RoHS
Protezione IP IP54 (splash and dust proof)
Potenza max 2W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.