Learn More
Mettler Toledo™ Nitrate Membrane Kit
Product Code. 10299681
Change view
Click to view available options
Material:
Polymer Membrane
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10299681 Polymer Membrane 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10299681 Supplier Mettler Toledo™ Supplier No. 51340011
  • ONLINE EXCLUSIVE PRICE
    740.00€
    620.69€ / 1 sets
    Risparmia 119.31€
    16% Sconto
In stock
Specifications
For Use With (Application) In gold Nitrate ISE
Material Polymer Membrane
Temperature (Metric) Operating 0°C to 50°C
pH Range 2 to 12