Learn More
Abcam Nitric Oxide Synthase Activity Assay Kit (Fluorometric)
Codice prodotto. 32118551 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
100 Tests
Dimensione della confezione:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32118551 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32118551 Fornitore Abcam N. del fornitore ab211084100Test
 Articolo soggetto a restrizioni
Spedizione stimata: 24-11-2026
Measure NOS activity in Cell/Tissue Extracts with Nitric Oxide Synthase Activity Assay Kit AB211084. Sensitivity = 0.75 μU. Cited in 4 publications. Use with Microplate reader.
Measure NOS activity in Cell/Tissue Extracts with Nitric Oxide Synthase Activity Assay Kit AB211084. Sensitivity = 0.75 μU. Cited in 4 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 100 Tests
Status giuridico RUO