Learn More
Parker Hannifin™ Nitrogen generator LCMS 30L with compressor 230V 50Hz Domnick Hunter
Product Code. 13263339
Change view
Click to view available options
Capacity:
30 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
13263339 30 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13263339 Supplier Parker Hannifin™ Supplier No. LCMS301E
  • ONLINE EXCLUSIVE PRICE
    34531.56€
    17140.91€ / pezzi 1
    Risparmia 17390.65€
    50% Sconto
Spedizione stimata: 22-03-2027
Specifications
Voltage 230 V
Electrical Requirements 230 V, 50 Hz
Includes Compressor
Capacity 30 L
Type Nitrogen Gas Generator