Learn More
Nor Doxepin Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | Nordoxepin Hydrochloride |
| CAS | 2887-91-4 |
| Note sul grado di purezza | HPLC |
| Formula molecolare | C18 H19 N O . HCl |
| Quantità | 25 mg |
| Sinonimo | 1-Propanamine','3-dibenz[b,e]oxepin-11(6H)-ylidene-N-methyl-','hydrochloride (1:1), 1-Propanamine','3-dibenz[b,e]oxepin-11(6H)-ylidene-N-methyl-','hydrochloride (9CI), Dibenz[b,e]oxepin-Δ11(6H),γ-propylamine','N-methyl-','hydrochloride (7CI,8CI), Dibenz[b,e]oxepin','1-propanamine deriv., Desmethyldoxepine hydrochloride, N-Desmethyldoxepin hydrochloride, Nordoxepin hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N-methylpropan-1-amine hydrochloride, Nordoxepin hydrochloride (cis/trans), N-Desmethyldoxepin hydrochloride (cis/trans), Doxepin-desmethyl hydrochloride |
| InChI Formula | InChI=1S/C18H19NO.ClH/c1-19-12-6-10-16-15-8-3-2-7-14(15)13-20-18-11-5-4-9-17(16)18;/h2-5,7-11,19H,6,12-13H2,1H3;1H |
| SMILES | Cl.CNCCC=C1c2ccccc2COc3ccccc13 |
| IUPAC Name | 3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N-methylpropan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 301.81 |
| Vedi altri risultati |