Learn More
uvex NORA S5 SRC Chemical-Resistant Safety Boots
Product Code. 15304183 Shop All Uvex Products
Change view
Click to view available options
Size:
38
40
43
45
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15304183 38 Each
15324183 40 Each
15354183 43 Each
15374183 45 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15304183 Supplier uvex Supplier No. 9475638
  • ONLINE EXCLUSIVE PRICE
    38.70€
    36.43€ / pezzi 1
    Risparmia 2.27€
    5% Sconto
Spedizione stimata: 09-12-2026
NORA safety boots are characterized by very high resistance to oils, grease, fuels, alkalis, acids and disinfectants making it the ideal boot choice when working with chemicals. Boots feature a robust protective steel toe cap, with a penetration-resistant steel midsole.
  • Boots are resistant to oils, grease, fuels, alkalis, acids, and disinfectants
  • Protection class S5 SRC in accordance with EN ISO 20345:2011
  • A robust, protective steel to cap with a penetration-resistant steel midsole
  • Free of plasticizers contained in the SVHC Candidate List of the REACH Regulation
  • Toe and heel reinforcement, as well as ankle protection
  • Anatomically shaped sole helps to prevent foot fatigue
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Material PVC
Size 38
Toe Material Steel
Gender Unisex
Resistant To Oils, grease, fuels, alkalis, acids, and disinfectants
Waterproof No
EN ISO 20345 S5
Product Type Safety Shoe