Learn More
Nordoxepin.HCl, Lipomed
Descrizione
Nordoxepin.HCl
Specifica
Specifica
| Materiale o nome chimico | Nordoxepin Hydrochloride |
| CAS | 2887-91-4 |
| Nomi componenti o analita | Nordoxepin Hydrochloride |
| Shipping Condition | Room Temperature |
| Formula molecolare | C18 H19 N O . HCl |
| Sinonimo | 1-Propanamine, 3-dibenz[b,e]oxepin-11(6 H)-ylidene-N-methyl-, hydrochloride (1:1), 1-Propanamine, 3-dibenz[b,e]oxepin-11(6 H)-ylidene-N-methyl-, hydrochloride (9 CI), Dibenz[b,e]oxepin-Δ11(6 H),γ-propylamine, N-methyl-, hydrochloride (7 CI,8 CI), Dibenz[b,e]oxepin, 1-propanamine deriv., Desmethyldoxepine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CNCCC=C1c2ccccc2COc3ccccc13 |
| IUPAC Name | 3-(6 H-benzo[c][1 ]benzoxepin-11-ylidene)-N-methylpropan-1-amine;hydrochloride |
| Industry Type | Pharmaceutical |
| Vedi altri risultati |