missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Novagen™ PopCulture™ Reagent

Formulated to extract proteins from E. coli cells directly in culture medium.

Marca:  Merck Millipore 71092-3

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 12868435

  • 90.26€ / 15mL


Spedizione stimata
01-02-2021
Aggiungi al carrello

Descrizione

Descrizione

  • Perforates E. coli cell wall without denaturing soluble protein protects protein from pH extremes
  • Assayed directly or purified by adding affinity matrix
  • Performed in single tube without need for columns or centrifugation
  • Applications:
    • High throughput (HT) robotic processing of samples for proteomics research
    • Packaging: Glass Bottle
Specifica

Specifica

15° to 30°C
15mL
Videos
SDS
Documenti