Learn More
Merck Chemicals Novagen™ Thrombin Cleavage Capture Kit
Descrizione
EMD Millipore Novagen™ Thrombin Cleavage Capture Kit used with any fusion protein that contains a thrombin recognition sequence.
- Convenient, quantitative removal of thrombin protease
- Monitors performance of cleavage conditions
- Easily visualized by SDS-PAGE
Specifica
Specifica
| Content And Storage | Multiple storage requirements |
| Tipo | Thrombin Cleavage Capture Kit |
| Quantità | 1 Kit |