Learn More
Invitrogen™ Novex™ OPD, 50 Tablets
Descrizione
- Used as substrate for horseradish peroxidase (HRP) in ELISAs
Build Your Own Immunoassay, Cell Analysis, Immunoassays
Order Info
Shipping condition: Wet ice
Specifica
Specifica
| Forma | Tablets |
| Materiale | Agarose |
| Quantità | 50 Tablets |