Learn More
NOX1 Antibody - BSA Free, Novus Biologicals™
Codice prodotto. 18663002 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.02 mL
0.1 mL
Dimensione della confezione:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18663002 0.02 mL 0.02mL
18676722 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18663002 Fornitore Novus Biologicals N. del fornitore NBP2947520.02ml
Rabbit Polyclonal Antibody
NOX1 Polyclonal antibody specifically detects NOX1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifica
Antigene NOX1
Applicazioni Western Blot
Classificazione Polyclonal
Coniugato Unconjugated
Diluizione Western Blot 1:100 - 1:500
Formulazione PBS (pH 7.3), 50% glycerol
Alias gene EC 1.6.3, GP91-2, Mitogenic oxidase 1, MOX-1, MOX1NADH/NADPH mitogenic oxidase subunit P65-MOX, NADPH oxidase 1, NADPH oxidase homolog-1, NOH1mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating), NOH-1NADPH oxidase 1 variant NOH-1L, NOX-1
Specie ospite Rabbit
Immunogeno Recombinant fusion protein containing a sequence corresponding to amino acids 415-564 of human NOX1 (NP_008983.2). KSIWYKFQCADHNLKTKKIYFYWICRETGAFSWFNNLLTSLEQEMEELGKVGFLNYRLFLTGWDSNIVGHAALNFDKATDIVTGLKQKTSFGRPMWDNEFSTIATSHPKSVVGVFLCGPRTLAKSLRKCCHRYSSLDPRKVQFYFNKENF
Metodo di purificazione Affinity purified
Quantità 0.02 mL
Status giuridico RUO
Disciplina di ricerca Cancer, Cell Biology, Direct Reversal of DNA Damage, Endocrinology, Immunology, Signal Transduction
Primario o secondario Primary
ID gene (immissione) 27035
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Forma Purified
Isotype IgG