missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Raccordi scanalati in PVDF

Combine these simple, economical PVDF male threaded adaptors with flexible tubing for use in your low-pressure applications (generally less than 30 psi). NPT Male Threaded Adapters slide into soft tubing. Tubing elasticity holds the tubing onto the fitting for low-pressure installations.

Marca:  Masterflex™ 30704-10

Dettagli aggiuntivi : Peso : 0.02400kg

 vedi altre versioni di questo prodotto

Codice prodotto. 11852613

  • 46.80€ / pezzi 10


In stock, pronto per la spedizione: 1
Aggiungi al carrello

Descrizione

Descrizione

  • Adapts from a NPT male thread to a hose barb
  • Designed for quick and easy assembly without tools
  • Excellent chemical resistance
  • Temp range: -62 to 129°C
  • Sterilize by ethylene oxide or autoclave
Specifica

Specifica

Barbed Fitting
Confezione da 10
1/8 NPT(M)
PDVF
1/8 NPT(M) thread to 3/8 in. tubing I.D.
3/31 in. orifice; 1-19/32 x 1/2 in.
Videos
SDS
Documenti