missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
CD73 (Ecto-5-prime-nucleotidase, 5-prime-ribonucleotide phosphohydrolase) catalyzes the conversion at neutral pH of purine 5-prime mononucleotides to nucleosides, the preferred substrate being AMP. CD73 consists of a dimer of 2 identical 70 kDa subunits bound externally to the plasma membrane by a glycosyl phosphatidyl inositol linkage. CD73 is used as a marker of lymphocyte differentiation. Consequently, a deficiency of CD73 occurs in a variety of immunodeficiency diseases. Other forms of 5-prime nucleotidase exist in the cytoplasm and lysosomes and can be distinguished from CD73 by their substrate affinities, requirement for divalent magnesium ion, activation by ATP, and inhibition by inorganic phosphate. The CD73 gene has been localized to 6q14-q21 by immunofluorescence and a study of a panel of human x mouse hybrids that contained fragments of chromosome 6 as translocations. Defects in the CD73 gene can lead to the calcification of joints and arteries, and intestinal tuberculosis. Two transcript variants encoding different isoforms of CD73 have been found.
Specifica
Specifica
| Antigene | NT5E/CD73 |
| Applicazioni | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classificazione | Polyclonal |
| Concentrazione | 0.24 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | NT5E |
| N. accesso geni | P21588, P21589, Q61503 |
| Alias gene | 5 NT, 5 nucleotidase, 5′ nucleotidase, ecto (CD73), CD73, E5NT, Ecto 5 nucleotidase, eN, eNT, NT, NT5, NT5E, NTE |
| Simboli geni | NT5E |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?