Learn More
Abnova™ NTRK3 (Texas Red/FITC) Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Marca:  Abnova™ FS0038.100uL
Dettagli aggiuntivi : Peso : 0.02000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 16063465
  • 1258.00€ / 100µL
Spedizione stimata
19-10-2020
Aggiungi al carrello
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Descrizione
Descrizione
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifica
Specifica
Fluorescent In Situ Hybridization (Cell)
Texas Red/FITC
DAPI Counterstain (1500 ng/mL ) 250 uL
Approx. 420kb/720kb
100 μL
Store at 4°C in the dark.
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Liquid
15q25/15q25
NTRK3 Split FISH Probe
Human
Genomic DNA