Learn More
Novus Biologicals™ NTS1/NTSR1 Overexpression Lysate
Codice prodotto. 18072928 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mg
Dimensione della confezione:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18072928 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18072928 Fornitore Novus Biologicals™ N. del fornitore NBP210833
Please to purchase this item. Need a web account? Register with us today!
The NTS1/NTSR1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifica
Linea cellulare The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_2522. The protein contains a C-terminal DDK tag.
Da utilizzare con (applicazione) Western Blot
Quantità 0.1 mg
Tampone di conservazione RIPA buffer
Purezza Protein
Test di specificità Homo sapiens Neurotensin Receptor 1, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native
For Research Use Only
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.