Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Columns
Product Code. 15559212
Change view
Click to view available options
Quantity:
50 Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15559212 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15559212 Supplier Macherey-Nagel™ Supplier No. 740609.50S
  • ONLINE EXCLUSIVE PRICE
    73.65€
    59.85€ / pezzi 50
    Risparmia 13.80€
    18% Sconto
In stock
Add to basket
Edge
Add to basket
Replacement mini spin columns for use with NucleoSpin™ Gel and PCR Clean-up kits to allow DNA fragments to be purified from enzymatic reactions, such as PCR, as well as from agarose gels.
NucleoSpin™ Gel and PCR Clean-up Kit — Two-in-one kit with optimized recovery and elution volume
  • Two applications in one kit – PCR clean-up and gel extraction
  • Uses one buffer with optimal performance for both applications
  • High recoveries for fragments down to 50 bp
  • Minimized elution volume of 15 μL – highly concentrated DNA
  • Binding buffer with pH indicator
  • Separate buffers for single stranded DNA or SDS containing samples available
  • Suitable for all gel buffer systems (e.g., TAE, TBE)
Specifications
Format Mini Spin Column
For Use With (Equipment) NucleoSpin Gel and PCR Clean-up Kit
Includes Silica Membrane
Product Type Spin Column
Quantity 50 Pk.
Product Identifier
  • NucleoSpin Gel and PCR Clean-up
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate