Learn More
Hamilton™ Nueros™ 33 Gauge Replacement Needles
Codice prodotto. 15655938
Cambia vista
Click to view available options
Calibro dell'ago:
33 G
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Calibro dell'ago
15655938 33 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15655938 Fornitore Hamilton™ N. del fornitore 6546102
  • ONLINE EXCLUSIVE PRICE
    234.68€
    222.27€ / pezzi 6
    Risparmia 12.41€
    5% Sconto
Spedizione stimata: 17-12-2026
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
  • Needle rigidity improves insertion path accuracy
  • Minimal tissue damage reduces injection variability
  • Reduced sample loss saves money and materials
  • Fine gauge needle creates smaller injection sites
  • Compatibility with most infusion pumps and stereotaxic holders means an easy integration into existing processes
Specifications
Autoclavabile Autoclavable
Calibro dell'ago 33 G
Lunghezza (sistema imperiale) ago 3.03 in.
Stile del punto dell'ago 4