Learn More
ACROBiosystems NY-ESO-1 specific TCR-HEK293 cell line
Codice prodotto. 31452722 Sfoglia Tutto ACROBiosystems Prodotti
Cambia vista
Click to view available options
Quantità:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
31452722 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 31452722 Fornitore ACROBiosystems N. del fornitore CHEKSTP1142Vials
 Articolo soggetto a restrizioni
Spedizione stimata: 31-08-2026
Aggiungi al carrello
Aggiungi al carrello
Cluster of Differentiation 3 (CD3) is a pivotal multimeric T cell co-receptor complex essential for adaptive immune responses.
Cluster of Differentiation 3 (CD3) is a pivotal multimeric T cell co-receptor complex essential for adaptive immune responses. Composed of epsilon (ε), gamma (γ), delta (δ), and zeta (ζ) polypeptide chains forming three distinct dimers (εγ, εδ, ζζ), CD3 non-covalently associates with the T-cell receptor (TCR) to establish the critical TCR/CD3 receptor complex on T cell surfaces. Structurally, CD3 proteins belong to the immunoglobulin superfamily, featuring a transmembrane domain, an N-terminal extracellular region with immunoglobulin-like domains, and cytoplasmic tails embedded with immunoreceptor tyrosine-based activation motifs (ITAMs). These ITAMs are fundamental for initiating signal transduction upon T cell receptor engagement, thereby activating both CD4+ T helper and CD8+ cytotoxic T cells. As a defining marker of the T cell lineage, CD3's central role in T cell activation makes its modulation, via stimulating or blocking antibodies, a promising therapeutic avenue for conditions like organ transplant rejection and autoimmune diseases.
Specifica
Quantità 2 Vials