Learn More
Brady™ Nylon Single Pole Circuit Breaker Lockout
Product Code. 11741635 Shop All Brady Products
Change view
Click to view available options
Color:
Red
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11741635 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11741635 Supplier Brady™ Supplier No. 065387
  • ONLINE EXCLUSIVE PRICE
    16.75€
    15.34€ / pezzi 1
    Risparmia 1.41€
    8% Sconto
In stock
Nylon Single Pole Circuit Breaker Lockout designed for 120 Volt breakers that have holes in the switch tongue. Quick and easy - just snap in place and apply lock. Dual cavities accommodate different size switch tongues.
  • Designed for 120 Volt breakers that have holes in the switch tongue.
  • Quick and easy - just snap in place and apply lock.
  • Dual cavities accommodate different size switch tongues.
  • Dimensions: 8.38 x 54.36 x 30.23 mm (D x W x H)
Specifications
Product Type Circuit Breaker Lockout
Color Red
Description Single Pole Circuit Breaker Lockout
Quantity 1 Ea.