missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Aminoazotoluene, TRC
Specifications
Specifications
| Materiale o nome chimico | 4-Amino-2,3-dimethylazobenzene |
| CAS | 97-56-3 |
| Formula molecolare | C14 H15 N3 |
| Quantità | 100 g |
| Sinonimo | Benzenamine, 2-methyl-4-[(2-methylphenyl)azo]- (9CI), C.I. Solvent Yellow 3 (8CI), o-Toluidine, 4-o-tolylazo- (6CI), 2-Methyl-4-[2-(2-methylphenyl)diazenyl]benzenamine, 2,3'-Dimethyl-4'-aminoazobenzene, 2-Aminoazotoluene, 2-Methyl-4-[(2-methylphenyl)diazenyl]amine, 2-Methyl-4-[(o-tolyl)azo]aniline, 2',3-Dimethyl-4-aminoazobenzene, 4-(o-Tolylazo)-o-toluidine, 4-Amino-2',3-dimethylazobenzene, Brasilazina Oil Yellow R, C.I. 11160, C.I. 11160B, Fast Garnet GBC Base, Fast Yellow AT, Fat Yellow B, Hidaco Oil Yellow, NSC 1797, NSC 26821, OAAT, Oil Yellow 21, Oil Yellow 2681, Oil Yellow 2R, Oil Yellow AT, Oil Yellow C, Oil Yellow I, Organol Yellow 2T, Solvent Yellow 3, Somalia Yellow R, o-AT, o-Aminoazotoluene |
| InChI Formula | InChI=1S/C14H15N3/c1-10-5-3-4-6-14(10)17-16-12-7-8-13(15)11(2)9-12/h3-9H,15H2,1-2H3 |
| SMILES | Cc1ccccc1N=Nc2ccc(N)c(C)c2 |
| IUPAC Name | 2-methyl-4-[(E)-(2-methylphenyl)diazenyl]aniline |
| Molecular Weight (g/mol) | 225.29 |
| Peso formulazione | 225.1266 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?