Learn More
DWK Life Sciences O-ring, made of nitrile, for safety standard ground sockets
Sfoglia Tutto DWK Life Sciences ProdottiDescrizione
Nitrile O-rings for DURAN™ safety ground joints together with the screw cap secure the connection between core and sleeve.
- In combination with the screw cap, the safety ground joints (core and sleeve) can be sealed tightly.
Specifica
Specifica
| Da utilizzare con (applicazione) | To secure the security grindings; Sealing |
| Joint Size | NS 45/40 |
| Quantità | 10/Pk. |
| Materiale | Nitrile |