missing translation for 'onlineSavingsMsg'
Learn More

OAS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 19279-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898944

  • 121.38€ / 20µL

Spedizione stimata: 04-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This antibody is specific to isoform p71 and isoform p69 of OAS2.

This gene encodes a member of the 2-5A synthetase family, essential proteins involved in the innate immune response to viral infection. The encoded protein is induced by interferons and uses adenosine triphosphate in 2′-specific nucleotidyl transfer reactions to synthesize 2′,5′-oligoadenylates. These molecules activate latent RNase L, which results in viral RNA degradation and the inhibition of viral replication. The three known members of this gene family are located in a cluster on chromosome 12. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifica

Specifica

OAS2
Polyclonal
Unconjugated
OAS2
(2 5)oligo(A) synthase 2, 2 5 oligoadenylate synthase 2, 2 5A synthase 2, OAS2, p69 OAS / p71 OAS, p69OAS / p71OAS
Rabbit
Antigen Affinity Chromatography
RUO
246728, 4939
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
E9Q9A9, P29728
OAS2
OAS2 Fusion Protein Ag6170
20 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali