Learn More
Obeticholic Acid Methyl Ester, Mikromol™
Descrizione
Obeticholic Acid Methyl Ester
Specifica
Specifica
| Materiale o nome chimico | Obeticholic Acid Methyl Ester |
| CAS | 951694-73-8 |
| Nomi componenti o analita | Obeticholic Acid Methyl Ester |
| Shipping Condition | Room Temperature |
| Formula molecolare | C27 H46 O4 |
| Certificazione/Conformità | ISO 17025 |
| Sinonimo | (3α,5β,6α,7α)-6-ethyl-3,7-dihydroxy-cholan-24-oic acid methyl ester, Methyl 3α,7-dihydroxy-6-ethyl-5β-cholanate, Obeticholic Acid Methyl Ester |
| InChI Formula | InChI=1S/C27H46O4/c1-6-18-22-15-17(28)11-13-27(22,4)21-12-14-26(3)19(16(2)7-10-23(29)31-5)8-9-20(26)24(21)25(18)30/h16-22,24-25,28,30H,6-15H2,1-5H3/t16-,17-,18-,19-,20+,21+,22+,24+,25-,26-,27-/m1/s1 |
| SMILES | CC[C@H]1[C@@H](O)[C@H]2[C@@H]3CC[C@H]([C@H](C)CCC(=O)OC)[C@@]3(C)CC[C@@H]2[C@@]4(C)CC[C@@H](O)C[C@@H]14 |
| IUPAC Name | methyl (4R)-4-[(3R,5S,6R,7R,8S,9S,10S,13R,14S,17R)-6-ethyl-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate |
| Vedi altri risultati |