Learn More
MedChemExpress Obinutuzumab
Specifica
Specifica
| Antigene | CD20 type II |
| Quantità | 1 mg |
| Target Species | Human |
| Content And Storage | Please store the product under the recommended conditions in the Certificate of Analysis. |
| Product Type | Inhibitory Antibodies |
| Shipping Condition | Room temperature |
| CAS | 949142-50-1 |
| Purity or Quality Grade | 99.4% |