Learn More
Justrite™ Oily Waste Can Foot Operated Cover & Soundgard
Codice prodotto. 12903565
Cambia vista
Click to view available options
Diametro (sistema imperiale) esterno:
13.94 in.
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Diametro (sistema imperiale) esterno Dimensione della confezione
12903565 13.94 in. Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 12903565 Fornitore Justrite™ N. del fornitore 09308
  • ONLINE EXCLUSIVE PRICE
    163.26€
    121.96€ / pezzi 1
    Risparmia 41.30€
    25% Sconto
Visualizza tutte le promozioni
Spedizione stimata: 29-12-2026
Foot operated for convenience and ease of use.
Rags and cloths soaked with solvents, thinners, linseed oil, combustible adhesives and other flammable liquids present a serious fire risk when improperly discarded. Lid opens to no more than 60 degrees and stays closed when not in use, isolating contents from fire sources and limiting oxygen to virtually eliminate the risk of spontaneous combustion.
Specifica
Diametro (sistema imperiale) esterno 13.94 in.
Lunghezza (sistema imperiale) 18.25 in.