Description
- Weight Sets in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
Specifications
| Certificazione/Conformità | NVLAP Accredited Certificate |
| Materiale | Stainless Steel, Aluminum |
| Peso (sistema metrico) | 0.050 kg |
| Massa | 500 mg to 1 mg |
| Include | Protective Case |
| Quantità confezione | 12/Pk. |