missing translation for 'onlineSavingsMsg'
Learn More

Oligomycin, TRC

Product Code. 30427635
Change view
Click to view available options
Quantità:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30427635 10 mg 10mg
30412163 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427635 Supplier Toronto Research Chemicals Supplier No. TRCO53297310MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico Oligomycin
CAS 1404-19-9
Formula molecolare C45H74O11
Quantità 10 mg
Sinonimo Oligomycin A','B','C mixture
InChI Formula InChI=1S/C45H74O11/c1-12-34-17-15-13-14-16-27(4)42(51)44(11,53)43(52)32(9)40(50)31(8)39(49)30(7)38(48)26(3)18-21-37(47)54-41-29(6)35(20-19-34)55-45(33(41)10)23-22-25(2)36(56-45)24-28(5)46/h13-15,17-18,21,25-36,38,40-42,46,48,50-51,53H,12,16,19-20,22-24H2,1-11H3/b14-13+,17-15+,21-18+/t25-,26-,27+,28+,29+,30-,31-,32-,33-,34-,35-,36-,38+,40+,41+,42-,44+,45-/m1/s1
SMILES CC[C@](CC[C@]1([H])[C@H](C)[C@]([C@@H](C)[C@]2(CC[C@@H](C)[C@](C[C@@H](O)C)([H])O2)O1)([H])OC3=O)([H])/C=C/C=C/C[C@H](C)[C@@H](O)[C@@](O)(C)C([C@H](C)[C@@H](O)[C@H](C)C([C@H](C)[C@@H](O)[C@H](C)/C=C/3)=O)=O
IUPAC Name (1S,4E,5'R,6R,6'R,7S,8R,10S,11S,12R,14S,15R,16S,18E,20E,22S,25R,27S,28R,29S)-22-ethyl-7,11,14,15-tetrahydroxy-6'-((S)-2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-3',4',5',6'-tetrahydro-2,26-dioxaspiro[bicyclo[23.3.1]nonacosane-27,2'-pyran]-4,18,20-triene-3,9,13-trione
Molecular Weight (g/mol) 791.06
Peso formulazione 790.52
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.