missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Olive Oil Acidity Test Kit with Magnetic Stirrer

Codice prodotto. 11686790
Cambia vista
Click to view available options
Quantità:
1 Kit
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11686790 1 Kit Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11686790 Fornitore Hanna Instruments™ N. del fornitore HI3897

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Utilizes a titration method where the endpoint is visually determined when the color changes from yellow-green to pink. Hanna Instruments™ Olive Oil Acidity Test Kit with Magnetic Stirrer is a user-friendly way to monitor and maintain the quality of olive oil at various stages of processing.

The percent acidity of olive oil is a direct indicator of its quality, classification, and freshness. Normally, testing acidity is a complicated process requiring various chemicals in a laboratory. The HI3897 is a perfect solution for olive oil producers who appreciate a speedy, accurate way to determine the quality of their product. With initial dissolution in an organic solvent, the olive oil sample is titrated with sodium hydroxide until a color change is observed. The titrant solution is added dropwise by a plastic calibrated syringe and tapered tip.

  • Single-Use Solvent Bottles: kit includes six bottles, each suitable for one titration test
  • Complete Setup: Kit includes all required equipment and solutions are included in the kit
  • Speedsafe™ Magnetic Stirrer: Capacity—1L, Speed—100 to 1000 rpm, ABS plastic cover
  • High Resolution Readings: 0.00 to 1.00% acidity determined to 0.01mL resolution
  • Replacement Reagents Available: A reagent kit containing titrant solution and organic solvent bottles for 10 tests can be ordered

Specifica

Accuracy 0.01 mL (0.01%) resolution
Descrizione Kit dimensions (LxWxH): 112 x 390 x 318mm (4.4 x 15.4 x 12.5 in.)
Stirrer dimensions (Dia. x H): 137mm x 51mm (5.39 x 2 in.)
Da utilizzare con (applicazione) Testing acidity level of olive oil
Include 6 bottles organic solvent with magnetic bar, magnetic stirrer, calibrated syringe for oil dosing, calibrated syringe for titrant dosing, titrant solution, carrying case, instructions
N. di test 6
Quantità 1 Kit
Range 0 to 1% acidity (as oleic acid)
Sufficiente per Sample size of 4.6mL / 4g
Metodo test Colorimetric
Tipo Olive Oil Acidity Test Kit
compliance-icons compliance-icons
Identificativo prodotto
  • TEST KIT ACIDITY OLIVE OIL
Avvertenza
  • Pericolo
Categoria di pericolo
  • Tossicità acuta Categoria 4
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Liquido infiammabile Categoria 2
  • Corrosione/irritazione della pelle Categoria 2
  • Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3
Indicazioni di pericolo
  • H225-Liquido e vapori facilmente infiammabili.
  • H302-Nocivo se ingerito.
  • H315-Provoca irritazione cutanea.
  • H319-Provoca grave irritazione oculare.
  • H336-Può provocare sonnolenza o vertigini.
  • EUH019-Può formare perossidi esplosivi.
  • EUH066-L'esposizione ripetuta può provocare secchezza o screpolature della pelle.
Consigli di prudenza
  • P210-Tenere lontano da fonti di calore, superfici riscaldate, scintille, fiamme e altre fonti di innesco. Vietato fumare.
  • P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
  • P312-Contattare un CENTRO ANTIVELENI/un medico/in caso di malessere.
  • P337+P313-Se l'irritazione degli occhi persiste, consultare un medico.
  • P370+P378-In caso d'incendio: utilizzare per estinguere.
  • P403+P235-Conservare in luogo fresco e ben ventilato.
Informazioni supplementari
  • MIXTURE LIST-Contain: Sodium hydroxide, Ethanol, Diethyl ether
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.