Learn More
EVOS™ Olympus Apochromat LWD Objective Lens
Codice prodotto. 15681882
Cambia vista
Click to view available options
Apertura numerica:
0.08
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Apertura numerica Dimensione della confezione
15681882 0.08 Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15681882 Fornitore EVOS™ N. del fornitore AMEP4751
Focus on Regenerative Therapies
Usa il codice promozionale 28027
Visualizza tutte le promozioni
Spedizione stimata: 14-10-2026
For demanding applications, especially capturing color images in white light
  • Has long working distance (LWD) objective that is optimized for imaging of slides, cell culture dishes and flasks, and microtiter plates
  • Compatible with EVOS™ Cell Imaging systems
  • Offers optical performance from visible light to near infrared light
  • Meets needs across spectrum of magnifications and optical specifications
TRUSTED_SUSTAINABILITY
Specifica
Tipo Olympus Apochromat LWD Objective Lens
Da utilizzare con (apparecchiatura) EVOS™ XL Core Imaging System, EVOS FL Color Imaging System, EVOS FL Auto Imaging System, EVOS XL Imaging System, EVOS FL Imaging System
Distanza focale 6.22mm
Ingrandimento 2X
Apertura numerica 0.08