missing translation for 'onlineSavingsMsg'
Learn More

Oncomodulin Antibody [DyLight 755], Novus Biologicals™

Codice prodotto. 30752504 Sfoglia Tutto Bio Techne Prodotti
Change view
Click to view available options
Quantità:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30752504 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30752504 Supplier Novus Biologicals Supplier No. AF6345Z

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Oncomodulin Polyclonal antibody specifically detects Oncomodulin in Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigene Oncomodulin
Applicazioni Western Blot
Classificazione Polyclonal
Coniugato DyLight 755
Formulazione 50mM Sodium Borate
Alias gene OCM1, OM, ONCM
Specie ospite Goat
Immunogeno E. coli-derived recombinant rat Oncomodulin, Met1-Ser109, Accession # P02631
Metodo di purificazione Antigen Affinity-purified
Quantità 0.1 mL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 654231
Target Species Rat
Content And Storage Store at 4°C in the dark.
Forma Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.