missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ONX-0914, MedChemExpress
Descrizione
ONX-0914 (PR-957) is a selective inhibitor of low-molecular mass polypeptide-7 (LMP7), the chymotrypsin-like subunit of the immunoproteasome. ONX-0914 blocks cytokine production and attenuates progression of experimental arthritis. ONX-0914 is a noncompetitive irreversible inhibitor of the mycobacterial proteasome (Ki=5.2 μM). ONX-0914 reactivates latent HIV-1 through p-TEFb activation mediated by HSF-1.
Specifica
Specifica
| Materiale o nome chimico | ONX-0914 |
| CAS | 960374-59-8 |
| Colore | White |
| Note sul grado di purezza | Research |
| Formula molecolare | C31H40N4O7 |
| Quantità | 50 mg |
| Sinonimo | PR-957 |
| Informazioni di solubilità | DMSO : ≥ 35 mg/mL (60.28 mM) |
| SMILES | O=C(N[C@@H](C)C(N[C@@H](CC1=CC=C(OC)C=C1)C(N[C@@H](CC2=CC=CC=C2)C([C@]3(C)OC3)=O)=O)=O)CN4CCOCC4 |
| Molecular Weight (g/mol) | 580.67 |
| Vedi altri risultati |
Research purposes only
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?