missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2S2 Polyclonal Antibody

Descrizione
This target displays homology in the following species: Cow: 93%; Dog: 79%; Guinea Pig: 85%; Horse: 86%; Human: 100%; Mouse: 79%; Pig: 86%; Rabbit: 79%; Rat: 85%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifica
Specifica
| Antigene | OR2S2 |
| Applicazioni | Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 0.5 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | OR2S2 |
| N. accesso geni | Q9NQN1 |
| Alias gene | olfactory receptor 2S2; olfactory receptor family 2 subfamily S member 2 (gene/pseudogene); olfactory receptor OR9-3; olfactory receptor, family 2, subfamily S, member 2; OR2S2; OR37A; OST715 |
| Simboli geni | OR2S2 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?