Learn More
Thermo Scientific™ Torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Gain improved plasma stability from the tulip design and easy, push-in Gain improved plasma stability from the tulip design and easy, push-in insertion of fully demountable, single piece, Thermo Scientific™ Torches with connection-free gas supply for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems. All connectivity (argon gas supplies and plasma ignition) is contained within the torch holder to reduce complexity and improve usability.
Specifica
Specifica
| Tipo | ICP-MS Quartz Torch |
| Da utilizzare con (apparecchiatura) | iCAP MX and iCAP Q/Qnova Series ICP-MS Systems |