Learn More
Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits
Codice prodotto. 15149192
Cambia vista
Click to view available options
Tipo:
Alkalinity
Aluminum
Ammonia
Bromine Reagent Tablet
Chloride
Chlorine
Chlorine Dioxide
Chlorine, Sodium Chlorate
Copper
Cyanuric Acid
Hydrazine
Iron
Manganese
Metallphthalein Method
Nitrate
Nitrite
Ozone
Phosphate
Silica
Sulfate
Sulfide
Zinc
pH
Range:
0.01 to 0.25 mg/L
0.01 to 0.3 mg/L Al
0.01 to 0.5 mg/L N
0.01 to 6 mg/L Cl2
0.02 to 1 mg/L Fe
0.02 to 1 mg/L N
0.02 to 1 mg/L Zn
0.02 to 11 mg/L ClO2
0.04 to 0.5 mg/L S
0.05 to 0.5 mg/L N2H4
0.05 to 0.5 mg/L O3
0.05 to 4 mg/L PO4
0.05 to 4 mg/L SiO2
0.05 to 5 mg/L Cu
0.1 to 13.5 mg/L
0.2 to 4 mg/L Mn
0.5 to 25 mg/L Cl-
1 to 40 mg/L
1 to 80 mg/L PO4
2 to 50 mg/L
5 to 100 mg/L SO4
5 to 200 mg/L
5 to 200 mg/L CaCO3
5 to 90 mg/L
6.5 to 8.4 pH
Quantità:
100 DPD No 1 Tablets
100 DPD No 4 Tablets
100 Tablets
100 Tests
30 Tests
50 Tests
50 Tests Each (free and total)
Dimensione della confezione:
pezzi 100
pezzi 30
pezzi 50
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo Range Quantità Dimensione della confezione
15149192 Chlorine, Sodium Chlorate 5 to 200 mg/L 100 Tests pezzi 100
11712238 Alkalinity 5 to 200 mg/L CaCO3 100 Tests pezzi 100
11782218 Aluminum 0.01 to 0.3 mg/L Al 50 Tests pezzi 50
15119192 Aluminum 0.01 to 0.25 mg/L 100 Tests pezzi 100
11752218 Ammonia 0.02 to 1 mg/L N 50 Tests pezzi 50
15109192 Bromine Reagent Tablet 0.1 to 13.5 mg/L - pezzi 100
11772218 Chloride 0.5 to 25 mg/L Cl- 50 Tests pezzi 50
11702248 Chlorine Dioxide 0.02 to 11 mg/L ClO2 100 Tests pezzi 100
11752238 Chlorine 0.01 to 6 mg/L Cl2 100 DPD No 1 Tablets pezzi 100
11732228 Chlorine 0.01 to 6 mg/L Cl2 50 Tests Each (free and total) pezzi 50
11762238 Chlorine 0.01 to 6 mg/L Cl2 100 DPD No 4 Tablets pezzi 100
11792218 Copper 0.05 to 5 mg/L Cu 50 Tests pezzi 50
11722228 Zinc 0.02 to 1 mg/L Zn 50 Tests pezzi 50
11792238 Cyanuric Acid 5 to 90 mg/L 100 Tests pezzi 100
15139192 Metallphthalein Method 2 to 50 mg/L 100 Tests pezzi 100
11782188 Hydrazine 0.05 to 0.5 mg/L N2H4 30 Tests pezzi 30
11772238 Iron 0.02 to 1 mg/L Fe 100 Tests pezzi 100
11702228 Manganese 0.2 to 4 mg/L Mn 50 Tests pezzi 50
11732218 Nitrate 1 to 40 mg/L 50 Tests pezzi 50
11722238 Nitrite 0.01 to 0.5 mg/L N 100 Tests pezzi 100
11732238 Ozone 0.05 to 0.5 mg/L O3 100 Tests pezzi 100
11702238 pH 6.5 to 8.4 pH 100 Tests pezzi 100
11742228 Phosphate 0.05 to 4 mg/L PO4 50 Tests pezzi 50
11752228 Phosphate 1 to 80 mg/L PO4 50 Tests pezzi 50
11712228 Silica 0.05 to 4 mg/L SiO2 50 Tests pezzi 50
11742238 Silica 0.05 to 4 mg/L SiO2 100 Tablets pezzi 100
11782238 Sulfate 5 to 100 mg/L SO4 100 Tests pezzi 100
11762218 Sulfide 0.04 to 0.5 mg/L S 100 Tests pezzi 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
Codice prodotto. 15149192 Fornitore Thermo Scientific™ N. del fornitore AC3072
  • ONLINE EXCLUSIVE PRICE
    89.95€
    74.52€ / pezzi 100
    Risparmia 15.43€
    17% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 27-11-2026
Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and other colorimeters
  • No. of tests: 50 to 100
TRUSTED_SUSTAINABILITY
Specifica
Tipo Chlorine, Sodium Chlorate
Da utilizzare con (apparecchiatura) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ3700 Colorimeter
Descrizione AQUAfast chlorine (total) HR & sodium chlorate reagent tablets, 100 tests
Range 5 to 200 mg/L
Quantità 100 Tests
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.