Learn More
Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes
Product Code. 11798236
Change view
Click to view available options
For Use With (Equipment):
Orion 810 Series DO Probes
Orion 830A Series DO Probes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11798236 Orion 810 Series DO Probes Each
11708246 Orion 830A Series DO Probes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11798236 Supplier Thermo Scientific™ Supplier No. 080160
  • ONLINE EXCLUSIVE PRICE
    189.00€
    176.00€ / pezzi 1
    Risparmia 13.00€
    6% Sconto
Spedizione stimata: 01-12-2026
Help ensure proper and optimal probe performance with the Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes.
Help ensure proper and optimal probe performance with Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes. Adapters include stir funnel and electrode adapter sleeve for BOD bottle.
Warranty: Out of box.
TRUSTED_SUSTAINABILITY
Specifications
Description BOD Adapter
For Use With (Equipment) Orion 810 Series DO Probes
Type BOD Adapter
Includes Stir funnel and electrode adaptor sleeve
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.