Learn More
Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes
Codice prodotto. 11732857
Cambia vista
Click to view available options
Tipo:
Epoxy Refillable Triode
Epoxy Triode Low Maintenance
Da utilizzare con (apparecchiatura):
Orion A+ Series Meters
Orion Star Series Meters
Dimensione della confezione:
Pezzo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo Da utilizzare con (apparecchiatura) Dimensione della confezione
11732857 Epoxy Triode Low Maintenance Orion Star Series Meters Pezzo
10691033 Epoxy Triode Low Maintenance Orion A+ Series Meters Pezzo
10315173 Epoxy Refillable Triode Orion A+ Series Meters Pezzo
10080514 Epoxy Refillable Triode Orion Star Series Meters Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 11732857 Fornitore Thermo Scientific™ N. del fornitore 9179BNMD
  • ONLINE EXCLUSIVE PRICE
    424.00€
    340.20€ / pezzi 1
    Risparmia 83.80€
    19% Sconto
In stock
Accurately measure ORP and temperature together in just one electrode, with a built-in temperature sensor.
Thermo Scientific™ Orion™ Triode 3-in-1 pH/Automatic Temperature Compensation Electrodes come with a built-in ATC and are engineered with an epoxy body for ruggedness. The 9179BN and 9179BNMD are low-maintenance gel-filled electrodes, best suited for educational institutes where minimal maintenance is required, while the 9180BNMD is a refillable electrode best suited for general purpose samples.
Orion™ Single Junction Electrodes provide a reliable option for routine ORP measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required:
Electrode fill solution for 9180BNMD
  • 900011 ORP filling solution for samples with total ionic strength > 0.2 M, 5 x 60 mL bottles
  • 900001 ORP filling solution for samples with total ionic strength < 0.2 M, 5 x 60 mL bottles
Electrode storage solution
  • 910001 4 M KCl storage solution, 475 mL bottle
Standards
  • 967901 ORP standard, 475 mL bottle
  • 967961 ORP standard, 5 x 60 mL bottles
pH cleaning solution kit
  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each
Items included with 9810BNMD
  • 900011 ORP filling solution for samples with total ionic strength > 0.2 M, 60 mL 1 bottle
Warranty period
  • 9179BN and 9179BNMD: 6 months from date of purchase
  • 9180BNMD: 12 months from date of purchase
TRUSTED_SUSTAINABILITY
Specifica
Applicazioni Water testing,wastewater, disinfection, aquaculture, process water.
Tipi di connettore BNC (Waterproof)
Descrizione Epoxy Triode Low Maintenance
Materiale Resina epossidica
Certificazione/Conformità ASTM D1498, Standard Methods 2580, USGS Ch 6.5
Da utilizzare con (apparecchiatura) Orion Star Series Meters
Temperatura (sistema metrico) operativa 0°C to 80°C
Tipo Epoxy Triode Low Maintenance