Learn More
Tocris Bioscience™ OSU 03012
PDPK1 (PDK1) inhibitor; inhibits Akt signaling
Marca: Tocris Bioscience™ 5682/50
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
OSU 03012 is a PDPK1 (PDK1) inhibitor; inhibits Akt signaling. Induces apoptosis of PC-3 and medulloblastoma cells, and inhibits growth of a number of tumor cell lines. Sensitizes radiotherapy-induced cell death and enhances cytotoxic effects of chemotherapeutic agents in vitro. Attenuates tumor growth of medulloblastoma xenografts in mice.Specifica
| 2-Amino-N-[4-[5-(2-phenanthrenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]phenyl]acetamide | |
| 742112-33-0 | |
| 50 mg |