Learn More
uvex OTG Plus Planet Clear SV EXC Overspecs, Anthracite/Jade
Codice prodotto. 31751169 Sfoglia Tutto Uvex Prodotti
Cambia vista
Click to view available options
Lens Material:
Polycarbonate
Dimensione della confezione:
pezzi 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Lens Material Dimensione della confezione
31751169 Polycarbonate pezzi 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 31751169 Fornitore uvex N. del fornitore 9139295
  • ONLINE EXCLUSIVE PRICE
    174.32€
    154.27€ / pezzi 8
    Risparmia 20.05€
    11% Sconto
Spedizione stimata: 11-12-2026
Provide optimum protection and high compatibility with prescription spectacles, featuring a large lens for excellent visibility and comfort.
  • Modern uvex OTG plus overspecs designed for prescription spectacle wearers, featuring an extremely large, flat lens that provides excellent visibility and sufficient space to comfortably wear even large prescription glasses underneath.
  • uvex protecting planet design with a lens made from mass-balanced polycarbonate, a frame partially manufactured from bio-based and recycled materials, plastic-free packaging, 100% recyclable construction, and completely metal-free design.
  • Innovative frame with multi-incline adjustment points allowing a high level of individual customisation to ensure a secure fit for all facial shapes and head widths.
  • Ergonomically shaped, flat flexible side arms with soft-grip ear-pieces deliver a comfortable, secure fit without pressure points, even during extended wear.
  • Anatomically designed nose piece ensures all-day wearing comfort and optimal compatibility with respirators.
  • Certified according to EN ISO 16321-1 for personal eye and face protection in industrial environments, including UV protection filtering.
  • Robust polycarbonate lens with proven uvex supravision coating technology providing permanent anti-fog performance and scratch resistance.
  • UV400 protection offering 100% shielding against hazardous ultraviolet radiation up to 400 nm.
TRUSTED_SUSTAINABILITY
Specifica
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Overglasses
Colore telaio Green
Materiale Polycarbonate
Monouso Reusable
Lens Material Polycarbonate
Materiale del telaio Polycarbonate
UV Protection UV400
Frame Type Adjustable Temples