Learn More
Cytiva Outlet Valve
Product Code. 15219774 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
AKTA start Automated Protein Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15219774 AKTA start Automated Protein Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15219774 Supplier Cytiva Supplier No. 29023898
In stock
For use with the AKTA start automated protein purification system. The outlet valve is used to direct the flow to the fraction collector or to the waste.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Direct the flow to the fraction collector or to the waste
For Use With (Equipment) AKTA start Automated Protein Purification System