Learn More
Thermo Scientific™ Owl™ Gel-Drying Kits
Codice prodotto. 11560435
Cambia vista
Click to view available options
Gel Size:
14 x 14 cm
24 x 24 cm
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Gel Size unitSize
11560435 24 x 24 cm Pezzo
12454637 14 x 14 cm Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11560435 Fornitore Thermo Scientific™ N. del fornitore GDF20
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures with these gel-drying kits.
Preserve protein gels easily and economically while eliminating distortion problems common in other drying procedures. Thermo Scientific™ Owl™ Gel-Drying Kits provide an inexpensive and simple way to produce a permanent record of gels by air drying them between two sheets of cellophane.
  • Available in minigel and large format sizes
  • Convenient method of preserving gels by air drying them between two sheets of cellophane
  • Beveled-edge design of gel-drying frame makes assembly easy
Includes:
Complete kits include two sets of gel-drying frames, 50 sheets of precut cellophane, two solid back plates, and eight side clips; frames and cellphane are also available separately
TRUSTED_SUSTAINABILITY
Specifica
Gel Size 24 x 24 cm
Quantità Each
Lunghezza (sistema metrico) 24 cm
Larghezza (sistema metrico) 24 cm
Product Type Gel Drying Kit